Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrnad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153562.4 | NP_705790.3 | 6 - 7 | 1356 | 64 | |
GenBank mRNA | AI592533.1 | - | 1 - 2 | 347 | 64 | |
AK035389.1 | - | 6 - 7 | 1308 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |