Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009796.2 | NP_033926.1 | 4 - 5 | 598 | 76 | |
GenBank mRNA | AB028640.1 | - | 4 - 5 | 598 | 76 | |
AJ012475.1 | - | 4 - 5 | 411 | 76 | ||
AK169334.1 | - | 4 - 5 | 643 | 76 | ||
BC095980.1 | - | 4 - 5 | 615 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |