Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162999.2 | NP_001156471.1 | 8 - 9 | 923 | 65 | |
GenBank mRNA | BC116316.1 | - | 1 - 2 | 187 | 65 | |
BC116317.1 | - | 1 - 2 | 187 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |