Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147201.2 | NP_671734.1 | 15 - 16 | 1483 | 79 | |
GenBank mRNA | AF302138.1 | - | 15 - 16 | 1482 | 79 | |
AK046142.1 | - | 16 - 17 | 1501 | 79 | ||
AK077440.1 | - | 15 - 16 | 1520 | 79 | ||
AK170501.1 | - | 15 - 16 | 1499 | 79 | ||
AK182568.1 | - | 4 - 5 | 271 | 79 | ||
AK185308.1 | - | 4 - 5 | 270 | 79 | ||
AK196002.1 | - | 4 - 5 | 270 | 79 | ||
AK215481.1 | - | 4 - 5 | 329 | 79 | ||
AK219460.1 | - | 4 - 5 | 263 | 79 | ||
BC004756.1 | - | 15 - 16 | 1453 | 3154089 | 79 | |
BC013212.1 | - | 4 - 5 | 877 | 79 | ||
BC018463.1 | - | 15 - 16 | 1457 | 3598438 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |