Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153680.2 | NP_710147.1 | 7 - 8 | 703 | 78 | |
GenBank mRNA | AK047265.1 | - | 6 - 7 | 797 | 78 | |
AK077650.1 | - | 7 - 8 | 705 | 78 | ||
AK077780.1 | - | 7 - 8 | 78 | |||
AK156299.1 | - | 7 - 8 | 754 | 78 | ||
AK219724.1 | - | 4 - 5 | 348 | 78 | ||
BC023732.1 | - | 7 - 8 | 719 | 5352355 | 78 | |
BC023925.1 | - | 2 - 3 | 166 | 78 | ||
BC026571.1 | - | 7 - 8 | 703 | 4986268 | 78 | |
BC029675.1 | - | 7 - 8 | 690 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |