Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007646.4 | NP_031672.2 | 6 - 7 | 813 | 84 | |
GenBank mRNA | AK038439.1 | - | 6 - 7 | 791 | 84 | |
AK040498.1 | - | 6 - 7 | 799 | 84 | ||
AK042970.1 | - | 6 - 7 | 782 | 84 | ||
AK150412.1 | - | 6 - 7 | 831 | 84 | ||
AK151016.1 | - | 6 - 7 | 825 | 84 | ||
AK151918.1 | - | 6 - 7 | 782 | 84 | ||
AK153215.1 | - | 6 - 7 | 829 | 84 | ||
AK153334.1 | - | 6 - 7 | 782 | 84 | ||
BC046312.1 | - | 6 - 7 | 789 | 6310453 | 84 | |
CT010386.1 | - | 6 - 7 | 764 | 84 | ||
L11332.1 | - | 6 - 7 | 770 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |