Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubgcp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198031.1 | NP_932148.1 | 10 - 11 | 1231 | 80 | |
GenBank mRNA | AK150149.1 | - | 10 - 11 | 1255 | 80 | |
AK165503.1 | - | 10 - 11 | 1254 | 80 | ||
AK171491.1 | - | 10 - 11 | 1237 | 80 | ||
AK171804.1 | - | 10 - 11 | 1246 | 80 | ||
BC025647.1 | - | 5 - 6 | 478 | 80 | ||
BC058566.1 | - | 10 - 11 | 1231 | 6466057 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |