Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gid8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289652.1 | NP_001276581.1 | 1 - 2 | 125 | 76 | |
GenBank mRNA | AK009106.1 | - | 1 - 2 | 85 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Detect Genomic DNA Mus musculus Probe spans exons |