Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181325.4 | NP_851842.1 | 2 - 3 | 295 | 63 | |
GenBank mRNA | AF133914.1 | - | 2 - 3 | 143 | 63 | |
AK052758.1 | - | 2 - 3 | 227 | 63 | ||
AK081243.1 | - | 2 - 3 | 218 | 63 | ||
AK142223.1 | - | 2 - 3 | 165 | 63 | ||
AK145966.1 | - | 2 - 3 | 165 | 63 | ||
AK154988.1 | - | 2 - 3 | 271 | 63 | ||
BC061143.1 | - | 2 - 3 | 145 | 30276598 | 63 | |
CJ172337.1 | - | 2 - 3 | 295 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons |