Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF093416.1 | - | 8 - 9 | 857 | 129 | |
AF099066.1 | - | 9 - 10 | 879 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |