Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ythdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177680.3 | NP_808348.2 | 10 - 11 | 1513 | 86 | |
GenBank mRNA | AB221619.1 | - | 2 - 3 | 137 | 86 | |
AI121974.1 | - | 3 - 4 | 248 | 86 | ||
AK040341.1 | - | 1 - 2 | 3630 | 86 | ||
AK173312.1 | - | 7 - 8 | 776 | 86 | ||
BC022697.1 | - | 3 - 4 | 306 | 86 | ||
BC037593.1 | - | 4 - 5 | 406 | 86 | ||
BC044650.1 | - | 8 - 9 | 1331 | 86 | ||
BC048817.1 | - | 3 - 4 | 291 | 86 | ||
CO424360.1 | - | 5 - 6 | 458 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |