Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007426.4 | NP_031452.2 | 7 - 8 | 1481 | 59 | |
GenBank mRNA | AF004326.1 | - | 7 - 8 | 1407 | 59 | |
AK019860.1 | - | 7 - 8 | 1470 | 59 | ||
AK048622.1 | - | 7 - 8 | 1438 | 59 | ||
AK143974.1 | - | 7 - 8 | 1469 | 59 | ||
AK156132.1 | - | 7 - 8 | 1452 | 59 | ||
BC027216.1 | - | 7 - 8 | 1434 | 3494566 | 59 | |
BC138309.1 | - | 7 - 8 | 1375 | 8861329 | 59 | |
BC138313.1 | - | 7 - 8 | 1375 | 8861333 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |