Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021375.3 | NP_067350.2 | 2 - 3 | 401 | 61 | |
GenBank mRNA | AF193808.1 | - | 2 - 3 | 401 | 61 | |
AK081882.1 | - | 2 - 3 | 402 | 61 | ||
AY254685.1 | - | 2 - 3 | 389 | 61 | ||
BC029236.1 | - | 2 - 3 | 419 | 4218889 | 61 | |
FJ387503.1 | - | 519 | 61 | |||
FJ387504.1 | - | 298 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |