Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007386.2 | NP_031412.2 | 17 - 18 | 2406 | 61 | |
GenBank mRNA | AK027914.1 | - | 17 - 18 | 2193 | 61 | |
AK085951.1 | - | 17 - 18 | 2156 | 61 | ||
AK129025.1 | - | 17 - 18 | 2188 | 61 | ||
AK163985.1 | - | 17 - 18 | 2205 | 61 | ||
BC005454.1 | - | 17 - 18 | 2166 | 3494686 | 61 | |
X61147.1 | - | 17 - 18 | 2173 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |