Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008628.2 | NP_032654.1 | 6 - 7 | 1115 | 67 | |
GenBank mRNA | AK051960.1 | - | 6 - 7 | 1112 | 67 | |
AK157842.1 | - | 6 - 7 | 1128 | 67 | ||
AK166179.1 | - | 6 - 7 | 1101 | 67 | ||
BC047117.1 | - | 6 - 7 | 1123 | 4951173 | 67 | |
BC050897.1 | - | 6 - 7 | 1098 | 6401835 | 67 | |
U21011.1 | - | 6 - 7 | 1132 | 67 | ||
X81143.1 | - | 6 - 7 | 1115 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |