Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcolce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029620.2 | NP_083896.1 | 2 - 3 | 352 | 70 | |
GenBank mRNA | AF352788.1 | - | 2 - 3 | 248 | 70 | |
AK010249.1 | - | 2 - 3 | 353 | 70 | ||
AK132395.1 | - | 242 | 70 | |||
BC051174.1 | - | 2 - 3 | 225 | 6310294 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |