Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159729.1 | NP_001153201.1 | 1 - 2 | 165 | 122 | |
GenBank mRNA | AK016493.1 | - | 1 - 2 | 177 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |