Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maged1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019791.2 | NP_062765.1 | 9 - 10 | 1982 | 71 | |
GenBank mRNA | AB029448.1 | - | 9 - 10 | 1982 | 71 | |
AF319975.1 | - | 8 - 9 | 1752 | 71 | ||
AK013231.1 | - | 2 - 3 | 201 | 71 | ||
AK017275.1 | - | 9 - 10 | 1900 | 71 | ||
AK049660.1 | - | 9 - 10 | 1933 | 71 | ||
AK087937.1 | - | 9 - 10 | 1932 | 71 | ||
AK132248.1 | - | 9 - 10 | 1921 | 71 | ||
AK135452.1 | - | 9 - 10 | 1932 | 71 | ||
AK145872.1 | - | 9 - 10 | 1902 | 71 | ||
AK146867.1 | - | 9 - 10 | 1932 | 71 | ||
AK146908.1 | - | 9 - 10 | 1932 | 71 | ||
AK150876.1 | - | 6 - 7 | 921 | 71 | ||
AK220150.1 | - | 10 - 11 | 1839 | 71 | ||
BC016438.1 | - | 8 - 9 | 2116 | 4480947 | 71 | |
BC031461.1 | - | 9 - 10 | 1922 | 4018875 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |