Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027382.3 | NP_081658.1 | 8 - 9 | 990 | 57 | |
GenBank mRNA | AK011332.1 | - | 8 - 9 | 991 | 57 | |
AK131998.1 | - | 8 - 9 | 1203 | 57 | ||
BC061257.1 | - | 8 - 9 | 962 | 30251620 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |