Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmtc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028651.2 | NP_082927.1 | 3 - 4 | 420 | 62 | |
GenBank mRNA | AK028333.1 | - | 3 - 4 | 401 | 62 | |
AK033947.1 | - | 3 - 4 | 421 | 62 | ||
AK036741.1 | - | 3 - 4 | 377 | 62 | ||
AK039093.1 | - | 3 - 4 | 365 | 62 | ||
AK043293.1 | - | 3 - 4 | 447 | 62 | ||
AK044391.1 | - | 3 - 4 | 376 | 62 | ||
AK045271.1 | - | 3 - 4 | 382 | 62 | ||
AK049748.1 | - | 4 - 5 | 561 | 62 | ||
AK082526.1 | - | 2 - 3 | 248 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |