Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172841.2 | NP_766429.2 | 1 - 2 | 1561 | 95 | |
GenBank mRNA | AK048882.1 | - | 1 - 2 | 1486 | 95 | |
BC117943.1 | - | 1 - 2 | 1340 | 40097907 | 95 | |
BC117944.1 | - | 1 - 2 | 1340 | 95 | ||
BQ031233.1 | - | 2 - 3 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |