Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172841.2 | NP_766429.2 | 3 - 4 | 1904 | 153 | |
GenBank mRNA | AK041736.1 | - | 3 - 4 | 1905 | 153 | |
BC117943.1 | - | 3 - 4 | 1683 | 40097907 | 153 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |