Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145452.1 | NP_001138924.1 | 12 - 13 | 1439 | 64 | |
GenBank mRNA | AK053944.1 | - | 8 - 9 | 1073 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Mus musculus Probe spans exons |