Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114361.1 | NP_001107833.1 | 16 - 17 | 2085 | 104 | |
NM_001114362.1 | NP_001107834.1 | 14 - 15 | 1878 | 104 | ||
NM_001286567.1 | NP_001273496.1 | 16 - 17 | 1988 | 104 | ||
NM_199466.3 | NP_955760.3 | 13 - 14 | 1749 | 104 | ||
GenBank mRNA | AK077527.1 | - | 8 - 9 | 913 | 104 | |
BC032258.1 | - | 2 - 3 | 233 | 104 | ||
BC065178.1 | - | 16 - 17 | 1926 | 6827397 | 104 | |
BC066099.1 | - | 13 - 14 | 1680 | 30533573 | 104 | |
BC067011.1 | - | 13 - 14 | 1698 | 6405184 | 104 | |
BC070427.1 | - | 14 - 15 | 1818 | 6413041 | 104 | |
BC079619.1 | - | 13 - 14 | 1680 | 30533573 | 104 | |
DQ230326.1 | - | 15 - 16 | 1768 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |