Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114361.1 | NP_001107833.1 | 9 - 10 | 1262 | 80 | |
NM_001114362.1 | NP_001107834.1 | 7 - 8 | 1055 | 80 | ||
NM_001286567.1 | NP_001273496.1 | 9 - 10 | 1165 | 80 | ||
NM_199466.3 | NP_955760.3 | 6 - 7 | 926 | 80 | ||
GenBank mRNA | AK077527.1 | - | 1 - 2 | 89 | 80 | |
AK144604.1 | - | 9 - 10 | 1263 | 80 | ||
BC065178.1 | - | 9 - 10 | 1103 | 6827397 | 80 | |
BC066099.1 | - | 6 - 7 | 857 | 30533573 | 80 | |
BC067011.1 | - | 6 - 7 | 875 | 6405184 | 80 | |
BC070427.1 | - | 7 - 8 | 995 | 6413041 | 80 | |
BC079619.1 | - | 6 - 7 | 857 | 30533573 | 80 | |
DQ230326.1 | - | 8 - 9 | 945 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |