Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008446.2 | NP_032472.2 | 6 - 7 | 1180 | 88 | |
GenBank mRNA | AK084837.1 | - | 6 - 7 | 1157 | 88 | |
AK135179.1 | - | 6 - 7 | 1128 | 88 | ||
AK135430.1 | - | 6 - 7 | 674 | 88 | ||
BC050946.1 | - | 6 - 7 | 1118 | 6822283 | 88 | |
BC054537.1 | - | 6 - 7 | 1102 | 88 | ||
D12646.1 | - | 6 - 7 | 1507 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |