Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npas4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153553.4 | NP_705781.1 | 3 - 4 | 588 | 73 | |
GenBank mRNA | AB049835.1 | - | 3 - 4 | 483 | 73 | |
AK032607.1 | - | 3 - 4 | 588 | 73 | ||
AY730724.1 | - | 3 - 4 | 615 | 73 | ||
BC059815.1 | - | 3 - 4 | 933 | 73 | ||
BC079544.1 | - | 3 - 4 | 933 | 73 | ||
BC129861.1 | - | 3 - 4 | 524 | 40110003 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |