Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080549.2 | NP_001074018.1 | 7 - 8 | 1853 | 88 | |
NM_001290459.1 | NP_001277388.1 | 6 - 7 | 1706 | 88 | ||
GenBank mRNA | AK083436.1 | - | 6 - 7 | 1694 | 88 | |
BC116300.1 | - | 7 - 8 | 1796 | 40095063 | 88 | |
BC116301.1 | - | 6 - 7 | 1649 | 40095064 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |