Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042779.2 | NP_001036244.1 | 10 - 11 | 1211 | 82 | |
NM_001291537.1 | NP_001278466.1 | 10 - 11 | 1475 | 82 | ||
NM_001291538.1 | NP_001278467.1 | 10 - 11 | 1469 | 82 | ||
NM_001291539.1 | NP_001278468.1 | 9 - 10 | 1152 | 82 | ||
NM_009153.3 | NP_033179.2 | 8 - 9 | 1021 | 82 | ||
NR_111986.1 | 8 - 9 | 1031 | 82 | |||
GenBank mRNA | AK014333.1 | - | 8 - 9 | 967 | 82 | |
AK033597.1 | - | 9 - 10 | 1162 | 82 | ||
AK037034.1 | - | 10 - 11 | 1200 | 82 | ||
AK043634.1 | - | 9 - 10 | 1105 | 82 | ||
AK048364.1 | - | 9 - 10 | 1433 | 82 | ||
AK148232.1 | - | 10 - 11 | 1172 | 82 | ||
BC090669.1 | - | 9 - 10 | 1132 | 30696359 | 82 | |
BC150758.1 | - | 7 - 8 | 867 | 9087669 | 82 | |
CN538654.1 | - | 1 - 2 | 67 | 82 | ||
X85990.1 | - | 8 - 9 | 1018 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |