Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011251.3 | NP_035381.2 | 10 - 11 | 2362 | 87 | |
NM_029169.3 | NP_083445.1 | 10 - 11 | 2633 | 87 | ||
GenBank mRNA | AJ006486.2 | - | 10 - 11 | 2241 | 87 | |
AK013915.1 | - | 1 - 2 | 55 | 87 | ||
AK032150.1 | - | 5 - 6 | 614 | 87 | ||
AK077899.1 | - | 10 - 11 | 2540 | 87 | ||
AK145612.1 | - | 10 - 11 | 2522 | 87 | ||
AK164432.1 | - | 5 - 6 | 719 | 87 | ||
AK220446.1 | - | 10 - 11 | 2511 | 87 | ||
BC026129.1 | - | 10 - 11 | 2387 | 3982861 | 87 | |
BC026130.1 | - | 10 - 11 | 2536 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |