Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190401.2 | NP_001177330.2 | 40 - 41 | 5507 | 83 | |
GenBank mRNA | CB520513.1 | - | 2 - 3 | 103 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |