Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271496.1 | NP_001258425.1 | 6 - 7 | 1156 | 126 | |
NM_013490.4 | NP_038518.2 | 5 - 6 | 1102 | 126 | ||
NR_073190.1 | 7 - 8 | 1216 | 126 | |||
GenBank mRNA | AB011002.1 | - | 5 - 6 | 961 | 126 | |
AK166465.1 | - | 5 - 6 | 1043 | 126 | ||
AK216139.1 | - | 2 - 3 | 209 | 126 | ||
BC004719.1 | - | 3 - 4 | 292 | 126 | ||
BC060218.1 | - | 5 - 6 | 1065 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |