Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081278.2 | NP_001074747.2 | 10 - 11 | 2783 | 73 | |
GenBank mRNA | AK031120.1 | - | 10 - 11 | 2627 | 73 | |
AK044719.1 | - | 10 - 11 | 2425 | 73 | ||
AK220347.1 | - | 9 - 10 | 1190 | 73 | ||
BC042747.1 | - | 6 - 7 | 785 | 73 | ||
BC064433.1 | - | 10 - 11 | 2407 | 73 | ||
BC080762.1 | - | 10 - 11 | 2401 | 73 | ||
BC117869.1 | - | 10 - 11 | 2256 | 40095526 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |