Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110504.1 | NP_001103974.1 | 5 - 6 | 686 | 64 | |
NM_007600.3 | NP_031626.1 | 5 - 6 | 738 | 64 | ||
GenBank mRNA | AF021847.1 | - | 5 - 6 | 617 | 64 | |
AF084459.1 | - | 4 - 5 | 604 | 64 | ||
AK149706.1 | - | 5 - 6 | 678 | 64 | ||
AK150084.1 | - | 5 - 6 | 677 | 64 | ||
AK160940.1 | - | 5 - 6 | 678 | 64 | ||
AK165258.1 | - | 5 - 6 | 697 | 64 | ||
AK168063.1 | - | 5 - 6 | 677 | 64 | ||
AK171405.1 | - | 5 - 6 | 641 | 64 | ||
AK196291.1 | - | 1 - 2 | 123 | 64 | ||
BC026138.1 | - | 5 - 6 | 653 | 4218148 | 64 | |
BC050276.1 | - | 5 - 6 | 654 | 4923347 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |