Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zic4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009576.2 | NP_033602.2 | 2 - 3 | 340 | 106 | |
GenBank mRNA | AK046913.1 | - | 2 - 3 | 341 | 106 | |
AK163477.1 | - | 2 - 3 | 1414 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |