Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm7sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028454.2 | NP_082730.2 | 1 - 2 | 183 | 61 | |
GenBank mRNA | AF480070.1 | - | 1 - 2 | 57 | 61 | |
AK014166.1 | - | 1 - 2 | 165 | 61 | ||
AK039028.1 | - | 1 - 2 | 113 | 61 | ||
AK040016.1 | - | 1 - 2 | 189 | 61 | ||
AK155475.1 | - | 1 - 2 | 140 | 61 | ||
BC014769.1 | - | 1 - 2 | 175 | 4239159 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |