Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm7sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028454.2 | NP_082730.2 | 5 - 6 | 732 | 86 | |
GenBank mRNA | AF480070.1 | - | 5 - 6 | 606 | 86 | |
AK014166.1 | - | 5 - 6 | 713 | 86 | ||
AK155475.1 | - | 5 - 6 | 689 | 86 | ||
BC014769.1 | - | 5 - 6 | 724 | 4239159 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |