Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053159.3 | NP_444389.2 | 5 - 6 | 722 | 94 | |
GenBank mRNA | AB049633.1 | - | 5 - 6 | 603 | 94 | |
AK008592.1 | - | 5 - 6 | 752 | 94 | ||
AK054185.1 | - | 2 - 3 | 157 | 94 | ||
AK077857.1 | - | 5 - 6 | 722 | 94 | ||
AK145687.1 | - | 5 - 6 | 752 | 94 | ||
AK169272.1 | - | 5 - 6 | 723 | 94 | ||
BC011128.1 | - | 5 - 6 | 691 | 4187019 | 94 | |
BC016586.1 | - | 5 - 6 | 595 | 2616397 | 94 | |
BC034877.1 | - | 5 - 6 | 688 | 1547459 | 94 | |
KC508086.1 | - | 5 - 6 | 699 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |