Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009767.2 | NP_033897.1 | 3 - 4 | 628 | 94 | |
GenBank mRNA | AK034427.1 | - | 3 - 4 | 601 | 94 | |
AK135974.1 | - | 3 - 4 | 580 | 94 | ||
BC132322.1 | - | 3 - 4 | 547 | 40130599 | 94 | |
BC138730.1 | - | 3 - 4 | 547 | 8861752 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |