Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001176.2 | NP_001001176.2 | 4 - 5 | 621 | 77 | |
NM_001167988.1 | NP_001161460.1 | 5 - 6 | 559 | 77 | ||
GenBank mRNA | AK131710.1 | - | 5 - 6 | 560 | 77 | |
AK135097.1 | - | 4 - 5 | 618 | 77 | ||
BC051635.1 | - | 4 - 5 | 419 | 77 | ||
BC066223.1 | - | 4 - 5 | 412 | 30468965 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |