Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700023214.mm.1 |
NM_001146222.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023214.mm.1 |
NM_001146222.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023214.mm.1 |
NM_001146223.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023214.mm.1 |
NM_001146223.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023214.mm.1 |
NM_001146224.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023214.mm.1 |
NM_001146224.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023214.mm.1 |
NM_024428.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023214.mm.1 |
NM_024428.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023214.mm.1 |
TC1700002534.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700023220.mm.1 |
NM_024428.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023220.mm.1 |
NM_024428.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700023220.mm.1 |
TC1700002534.mm.1 |
MTA TC |