Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027864.2 | NP_082140.2 | 2 - 3 | 697 | 82 | |
GenBank mRNA | AB175681.1 | - | 2 - 3 | 305 | 82 | |
AK002747.1 | - | 2 - 3 | 669 | 82 | ||
AK049598.1 | - | 2 - 3 | 643 | 82 | ||
AK078292.1 | - | 2 - 3 | 310 | 82 | ||
BC117800.1 | - | 2 - 3 | 481 | 40092267 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |