Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029402.3 | NP_083678.2 | 5 - 6 | 619 | 120 | |
GenBank mRNA | AK004875.2 | - | 5 - 6 | 575 | 120 | |
AK016520.1 | - | 5 - 6 | 589 | 120 | ||
AK144693.1 | - | 5 - 6 | 592 | 120 | ||
AK160597.1 | - | 5 - 6 | 604 | 120 | ||
AK220287.1 | - | 5 - 6 | 594 | 120 | ||
BC026779.1 | - | 5 - 6 | 520 | 4018298 | 120 | |
BC027428.1 | - | 5 - 6 | 582 | 3983461 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |