Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naaladl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009546.1 | NP_001009546.1 | 2 - 3 | 373 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Detect Genomic DNA Mus musculus Probe spans exons |