Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naaladl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009546.1 | NP_001009546.1 | 8 - 9 | 1213 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mus musculus Probe spans exons |