Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000010247.mm.1 |
NM_001162904.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010247.mm.1 |
NM_001162904.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010247.mm.1 |
NM_001162905.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010247.mm.1 |
NM_001162905.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010247.mm.1 |
NM_010785.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010247.mm.1 |
NM_010785.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010247.mm.1 |
NM_148922.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010247.mm.1 |
NM_148922.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010247.mm.1 |
TC1000001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000010248.mm.1 |
NM_001162904.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010248.mm.1 |
NM_001162904.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010248.mm.1 |
NM_001162905.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010248.mm.1 |
NM_001162905.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010248.mm.1 |
NM_010785.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010248.mm.1 |
NM_010785.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010248.mm.1 |
NM_148922.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010248.mm.1 |
NM_148922.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010248.mm.1 |
TC1000001374.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000010246.mm.1 |
NM_001162904.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010246.mm.1 |
NM_001162904.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010246.mm.1 |
NM_001162905.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010246.mm.1 |
NM_001162905.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010246.mm.1 |
NM_010785.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010246.mm.1 |
NM_010785.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010246.mm.1 |
NM_148922.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010246.mm.1 |
NM_148922.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010246.mm.1 |
TC1000001374.mm.1 |
MTA TC |