Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pof1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181579.1 | NP_853557.1 | 2 - 3 | 398 | 104 | |
GenBank mRNA | AF309775.1 | - | 1 - 2 | 291 | 104 | |
AK036922.1 | - | 2 - 3 | 403 | 104 | ||
AK086511.1 | - | 2 - 3 | 104 | |||
AK137653.1 | - | 2 - 3 | 399 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |