Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010401.3 | NP_034531.1 | 18 - 19 | 2130 | 91 | |
GenBank mRNA | AK014518.1 | - | 18 - 19 | 2130 | 91 | |
AK017397.1 | - | 3 - 4 | 304 | 91 | ||
AK028963.1 | - | 18 - 19 | 1837 | 91 | ||
AK081409.1 | - | 18 - 19 | 1833 | 91 | ||
BC057637.1 | - | 18 - 19 | 1807 | 4211545 | 91 | |
BC157999.1 | - | 18 - 19 | 2112 | 9088147 | 91 | |
L07645.1 | - | 18 - 19 | 1814 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |