Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prepl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163622.1 | NP_001157094.1 | 6 - 7 | 1232 | 76 | |
NM_001163623.1 | NP_001157095.1 | 5 - 6 | 696 | 76 | ||
NM_001163624.1 | NP_001157096.1 | 5 - 6 | 564 | 76 | ||
NM_145984.3 | NP_666096.3 | 6 - 7 | 796 | 76 | ||
GenBank mRNA | AK028877.1 | - | 6 - 7 | 1190 | 76 | |
AK050878.1 | - | 5 - 6 | 558 | 76 | ||
AK083070.1 | - | 6 - 7 | 737 | 76 | ||
BC004612.1 | - | 5 - 6 | 650 | 3585402 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |