Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008973.2 | NP_001008973.2 | 10 - 11 | 1226 | 90 | |
NM_001289485.1 | NP_001276414.1 | 11 - 12 | 1311 | 90 | ||
NM_001289486.1 | NP_001276415.1 | 10 - 11 | 1223 | 90 | ||
GenBank mRNA | AF338468.1 | - | 8 - 9 | 990 | 90 | |
AF338469.1 | - | 11 - 12 | 1305 | 90 | ||
AK053319.1 | - | 4 - 5 | 560 | 90 | ||
AK135727.1 | - | 2 - 3 | 217 | 90 | ||
BC050244.1 | - | 8 - 9 | 1011 | 90 | ||
BC150885.1 | - | 10 - 11 | 1188 | 9087796 | 90 | |
BC172032.1 | - | 10 - 11 | 1185 | 9054318 | 90 | |
DV662151.1 | - | 2 - 3 | 186 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |